Learn More
Medchemexpress LLC Suc-Ile-Glu(γ-pip)-Gly-Arg-pNA hydrochloride | 1379822-04-4 | 98.7% | 797.30 | 10 MG

Supplier: Medchemexpress LLC HYP312610MG
Suc-Ile-Glu(γ-pip)-Gly-Arg-pNA hydrochloride is a factor Xa specific chromogenic substrate.
- High purity of 98.7%
- Available as a white to off-white solid
- Powder storage: -80°C for 2 years; -20°C for 1 year
- In solvent storage: -80°C for 6 months; -20°C for 1 month
- Sealed storage, away from moisture and light, under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.