Learn More
Medchemexpress LLC Suc-Ile-Glu(γ-pip)-Gly-Arg-pNA hydrochloride | 1379822-04-4 | 98.7% | 797.30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP31265MG
Encompass_Preferred
Suc-Ile-Glu(γ-pip)-Gly-Arg-pNA hydrochloride is a factor Xa specific chromogenic substrate, intended for research use only. It has a purity of 98.68% and a molecular weight of 797.30.
  • Factor Xa specific chromogenic substrate
Catalog No. 50-003-97380