Learn More
Aat Bioquest (Suc-LLVY)2R110
SDP
Supplier:  Aat Bioquest 13451
Encompass_Preferred
(Suc-LLVY)2R110 is a sentive fluorogenic substrate for 20S proteasome, calpains and other chymotrypsin-like proteases.
Catalog No. 50-195-3449