Learn More
Carbosynth LLC. Suc-Phe-Ala-Ala-Phe-pNA, 200mg, CAS# 110906-89-3, MF= C34H38N6O9
SDP
Supplier:  Carbosynth LLC. SUB3007PI200MG
Encompass_Preferred
Application: Subtilisin and Myroilysin Substrate, salt: Trifluoroacetate Salt, one letter code: Suc-FAAF-pNA, MW: 674.72
Catalog No. 50-195-2434