Learn More
CPC Scientific Suc-Tyr-Val-Ala-Asp-AMC 1MG
SDP
Supplier:  CPC Scientific CASP030A
Encompass
Fluorogenic substrate for caspase-1.ONE-LETTER SEQUENCE: Suc-YVAD-AMCMOLECULAR FORMULA: C35H41N5O12MOLECULAR WEIGHT:723.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [201860-29-9], RESEARCH AREA: Apoptosis
Catalog No. 50-210-8976