Learn More
Cayman Chemical Suc-YVAD-AMCacetate 1mg
SDP
Supplier:  Cayman Chemical 271381 MG
Encompass_Preferred
A fluorogenic substrate for caspase-1; upon enzymatic cleavage, AMC is released and its fluorescence can be used to quantify caspase-1 activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-294-0359