Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Succinic acid-13C2 | 79864-95-2 | 120.07 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN0420S35MG

Encompass_Preferred

Butanedioic acid-13C2 is the 13C labeled Succinic acid. Succinic acid is an intermediate product of the tricarboxylic acid cycle and can be used as a precursor for various industrially important chemicals in food, chemical, and pharmaceutical industries. It is also described as a potent and orally active anxiolytic agent. This product is for research use only and not for sale to patients.

  • Butanedioic acid-13C2 is the 13C labeled succinic acid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.0%.
  • Appearance: Solid, white to off-white.

Catalog No. 50-003-89985


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.