Learn More
Medchemexpress LLC Succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate | 64987-85-5 | MFCD00009634 | 99.6% | 334.32 g/mol | C16H18N2O6 | 100 G
SDP
Supplier:  Medchemexpress LLC HY42360100G
Encompass_Preferred
Succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate is a heterobifunctional, non-cleavable crosslinker that couples primary amines to sulfhydryl groups via NHS-ester and maleimide chemistry. It is used to prepare stable protein-protein and protein-small molecule conjugates for applications such as antibody-drug conjugates and antigen conjugates for immunization.
  • Heterobifunctional, non-cleavable crosslinker
  • Reacts with primary amines (NHS ester) and sulfhydryls (maleimide)
  • Used for protein conjugation and antibody-drug conjugate synthesis
  • High purity suitable for research applications
  • Store sealed, protect from moisture and light; recommended cold storage
Catalog No. 50-002-03360