Learn More
Medchemexpress LLC HY-14937 5mg Medchemexpress, Succinobucol CAS:216167-82-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY149375MG
Encompass_Preferred
Medchemexpress, HY-14937 5mg Succinobucol CAS:216167-82-7 Succinobucol is a phenolic antioxidant with anti-inflammatory and antiplatelet effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1663