Learn More
Alkali Scientific Succinyl Chloride, 1 Kilogram
SDP
Supplier:  Alkali Scientific UCC001CP
Encompass_Preferred
Succinyl Chloride is an acid chloride used as a reagent in organic synthesis. It is highly reactive and is commonly employed in the production of succinyl derivatives, which are used in pharmaceutical, chemical, and biotechnological applications. Succinyl chloride is particularly important in the synthesis of peptides and other bioactive compounds, serving as an acylating agent in peptide synthesis and enzyme modification. This 1-kilogram quantity is suitable for large-scale chemical processes and research projects. It is essential for the production of intermediates used in pharmaceuticals, cosmetics, and agrochemicals.
Catalog No. 50-006-60537