Learn More
Medchemexpress LLC Succinylsulfathiazole | 116-43-8 | 99.22% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB092150MG
Encompass_Preferred
Succinylsulfathiazole is a long-acting, orally active sulfonamide antibiotic with localized gut-specific antibacterial activity. It functions by inhibiting bacterial folate synthesis, which in turn reduces coliform counts and suppresses intestinal bacterial growth and vitamin biosynthesis. Additionally, it modulates hepatic mTOR signaling, decreases cecal fermentation, reduces hepatic folate levels, and diminishes the fermentability of certain dietary fibers.
  • Long-acting sulfonamide antibiotic
  • Orally active with localized gut-specific antibacterial activity
  • Inhibits bacterial folate synthesis
  • Reduces coliform counts and suppresses intestinal bacterial growth
  • Modulates hepatic mTOR signaling
  • Decreases cecal fermentation and reduces hepatic folate levels
  • Can induce folate deficiency in some models
Catalog No. 50-004-09237