Learn More
Medchemexpress LLC HY-B0644 100mg Medchemexpress, Sucralfate CAS:54182-58-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0644100MG
Encompass_Preferred
Medchemexpress, HY-B0644 100mg Sucralfate CAS:54182-58-0 Sucralfate (Sucrose octasulfate-aluminum complex) is a potent and orally active gastroprotectant with no systemic effects. Sucralfate inhibits peptic activity and binds to negatively charged subepithelial proteins exposed during mucosal injury, forming a viscous layer that protects the vascular bed and proliferative zone. Sucralfate is used for prevention and treatment of several gastrointestinal diseases in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2630