Learn More
Medchemexpress LLC Sucralose-d6 | 1459161-55-7 | 403.67 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0614S1MG
Encompass_Preferred
Sucralose-d6 is deuterium labeled Sucralose. Sucralose (E955; Trichlorosucrose) is a non-nutritive artificial sweetener and sugar substitute. Sucralose can activate a conserved neural fasting response and thereby exerts an appetite-stimulating effect in rodents.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88972