Learn More
U.S. Pharmacopeia Sucrose, 57-50-1, MFCD00006626, 100 mg
Supplier:  U.S. Pharmacopeia 1623637
Encompass
Empirical Formula (Hill Notation): C12H22O11, Molecular Weight: 342.30, MP: 185-187 °C, Synonym: α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar.
Catalog No. NC9595525