Learn More
Medchemexpress LLC Sucrose-d14 (D-(+)-Saccharose-d14) | 57-50-1 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1779S31MG
Encompass_Preferred
Sucrose-d14 is the deuterium labeled Sucrose, a disaccharide composed of two monosaccharides, glucose and fructose. It can be applied in animal models for metabolic disease, obesity, and diet preference. This product serves as a valuable tool for quantitative analysis in various research applications.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
Catalog No. 50-004-11135