Learn More
BURKLE INC Suction head, suction drill 0.16 in. (4 mm), Mini ViscoSampler
Supplier:  BURKLE INC 53411004
Encompass_Preferred
Depending on the viscosity, the suction tip can be supplied with a suction hole of 2 mm to 6 mm. Compatible with Burkle Mini Visco Sampler (sold separately).
Catalog No. 50-000-25665