Learn More
Sklar Instruments Suction Tube Cleaning Brush, Sklar, 2mm x 8 in. Suction Tube Cleaning Brush For Baron and Frazier Suction Tubes, Nylon / Stainless Steel. PK/3.
SDP
Supplier:  Sklar Instruments 101400
Encompass_Preferred
Suction Tube Cleaning Brush, Sklar, 2mm x 8 in. Suction Tube Cleaning Brush For Baron and Frazier Suction Tubes, Nylon / Stainless Steel. Sklar's Suction Tube Cleaning Brushes have been specifically designed for use with Baron and Frazier suction tubes. They are available in a number of narrow brush tips to accommodate the various size Baron and Frazier tubes. This product has a 2mm wide brush tip and an overall length of 8 inches. They are sold in packs of three.
Catalog No. 50-143-6975