Learn More
Apexbio Technology LLC Sudan I 842-07-9 500mg
SDP
Supplier:  Apexbio Technology LLC C7176500
Encompass_Preferred
Sudan I (842-07-9) is a small-molecule inhibitor targeting microbial growth notably Clostridium perfringens and Clostridium ramosum It is used to modulate bacterial physiology by altering signaling pathways involved in cell viability and replication Sudan I exerts its biological activity primarily through disruption of cellular metabolic processes essential for microbial proliferation In in vitro studies Sudan I demonstrates inhibitory activity with IC50 values typically between 10 30 M depending on the bacterial strain and experimental conditions Based on these pharmacological properties Sudan I holds research potential in microbiology investigation and antimicrobial assay development particularly for studies on microbial inhibition mechanisms and comparative efficacy testing
Catalog No. 50-000-90280