Learn More
Medchemexpress LLC Sudan I-d5 | 752211-63-5 | 99.0% | 253.31 g/mol | C16H7D5N2O | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW0197761MG
Encompass_Preferred
Sudan I-d5 is the deuterated isotopologue of Sudan I (Solvent Yellow 14) supplied as an analytical reference standard for laboratory use. It is intended for use as a stable-isotope internal standard to support accurate quantitation, method validation, and peak identification in chromatographic and mass-spectrometric assays.
  • Suitable as an internal standard for LC-MS and GC-MS analyses.
  • Provides isotopic labeling for improved quantitation and peak identification.
  • High chemical purity for analytical applications.
  • Supplied in a small pack size for trace-level calibration and method development.
  • Applicable to food, environmental, and contaminant analysis workflows.
Catalog No. 50-003-00253