Learn More
Medchemexpress LLC Sudan I-d5 | 752211-63-5 | 99.0% | 253.31 g/mol | C16H7D5N2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW01977610MG
Encompass_Preferred
Sudan I-d5 is the deuterium-labeled (d5) stable-isotope analogue of Sudan I used as an analytical standard for chromatography and mass spectrometry. It serves as an internal standard for quantitative assays, method development, and trace-level detection of Sudan I in oils, food matrices, and environmental samples.
  • Deuterium-labeled internal standard for LC-MS/MS and GC-MS.
  • High isotopic labeling for reliable peak identification.
  • 99.0% chemical purity to minimize matrix interference.
  • Soluble in DMSO; warming and sonication improve dissolution.
  • Stable as powder at -20°C for long-term storage.
Catalog No. 50-002-99699