Learn More
Medchemexpress LLC Sudan I (Solvent Yellow 14) | 842-07-9 | 99.8% | 50 G
SDP
Supplier:  Medchemexpress LLC HYD002450G
Encompass_Preferred
Sudan I (Solvent Yellow 14) is a diazo-conjugate red dye commonly used as an additive in various products like oils, solvents, or polishes. This monoazo dye has the chemical formula of 1-phenylazo-2-naphthol.
  • Inhibits the growth of bacterial strains such as Clostridium perfringens and L. rhamnosus.
  • Potential mutagen and carcinogen for humans.
  • Induces genotoxic effects in HepG2 cells.
  • Increases mutagenic and clastogenic effects in MCL-5 cell lines.
  • Shows positive results in Salmonella Typhimurium mutagenicity tests with S9 activation.
Catalog No. 50-004-10793