Learn More
Medchemexpress LLC Sudan III-d6 | 1014689-17-8 | 93.0% | 1 MG
Supplier: Medchemexpress LLC HYD0931S1MG
Sudan III-d6 is a deuterium labeled Sudan III, which is a hydrophobic bisazo dye. Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development, and deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS