Learn More
Medchemexpress LLC Sugammadex sodium | 343306-79-6 | 98.0% | C72H104Na8O48S8 | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB0079200MG
Encompass_Preferred
Sugammadex sodium is a synthetic γ-cyclodextrin derivative that acts as a reversal agent for neuromuscular block. It also shows a nephroprotective effect in ischemia-reperfusion injury.
  • Acts as a reversal agent for neuromuscular block.
  • Exhibits nephroprotective effects in ischemia-reperfusion injury.
Catalog No. 50-004-32544