Learn More
Apexbio Technology LLC Sulconazole Nitrate 61318-91-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B20365.1
Encompass_Preferred
Sulconazole Nitrate (CAS 61318-91-0) is a small-molecule inhibitor targeting the fungal cytochrome P450-dependent enzyme lanosterol 14 -demethylase It is designed to inhibit this enzyme thereby disrupting ergosterol biosynthesis and compromising fungal cell membrane integrity Sulconazole Nitrate exerts its biological activity primarily through inhibition of lanosterol 14 -demethylase In in vitro studies Sulconazole Nitrate demonstrates broad-spectrum antifungal inhibition with IC50 values generally ranging between 1 to 10 M depending on fungal species and assay methods Based on these pharmacological properties Sulconazole Nitrate holds research potential in the evaluation of antifungal susceptibility profiles and the study of fungal membrane-associated targets and resistance mechanisms
Catalog No. 50-000-92854