Learn More
Medchemexpress LLC Sulfabrom | 116-45-0 | 357.23 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYU0013110MG
Encompass_Preferred
Sulfabrom is a long-acting sulfonamide used for treating coccidiosis and various bacterial infections in poultry, swine, and cattle. This compound is intended for research use only.
  • Long-acting sulfonamide
  • Used in veterinary applications
  • High purity of 99.0%
  • Molecular weight of 357.23
  • Appears as a white to light yellow solid
Catalog No. 50-004-48549