Learn More
Medchemexpress LLC Sulfabrom | 116-45-0 | 357.23 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYU0013110MM/1ML
Encompass_Preferred
Sulfabrom is a long-acting sulfonamide. It is used for the treatment of coccidiosis and various bacterial infections in poultry, swine, and cattle.
  • Used for treating coccidiosis
  • Effective against various bacterial infections
  • Applicable in poultry, swine, and cattle
Catalog No. 50-004-48510