Learn More
Medchemexpress LLC Sulfabrom | 116-45-0 | 357.23 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYU00131
Encompass_Preferred
Sulfabrom is a long-acting Sulfonamide (N 3517; Sulfabromomethazine) primarily used for the treatment of coccidiosis and various bacterial infections in poultry, swine, and cattle. This product is intended for research use only and is not sold for patient use.
  • Long-acting sulfonamide
  • Treats coccidiosis
  • Effective against various bacterial infections
  • Suitable for poultry, swine, and cattle
  • For research use only
Catalog No. 50-004-49423