Learn More
Medchemexpress LLC Sulfacetamide | 144-80-9 | 1 G
SDP
Supplier:  Medchemexpress LLC HYN71231G
Encompass_Preferred
Sulfacetamide (Sulphacetamide) is a sulfonamide antibiotic with antifungal and antibacterial activities. It is used as a topical formulation in various ophthalmic, dermatological, and hair solutions for bacterial infections, acne, and scalp conditions. Sulfacetamide targets bacterial folic acid synthesis and is effective against various gram-positive and gram-negative bacteria. It inhibits the synthesis of both dihydro-folic acid and para-aminobenzoic acid. It can be used for the study of ocular infections.
  • Can be used for the study of various ocular infections like trachoma, blepharitis, conjunctivitis, corneal ulcer, pityriasis versicolor and rosacea.
  • It is an antifungal drug through a CYP51A1-independent mechanism.
  • It is a bacteriostatic agent with similar activity against both gram-positive and gram-negative bacteria.
Catalog No. 50-004-17819