Learn More
Medchemexpress LLC Sulfacetamide | 144-80-9 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HYN712310MM/1ML
Encompass_Preferred
Sulfacetamide is a sulfonamide antibiotic used in topical formulations for ophthalmic, dermatological, and hair solutions. It targets bacterial folic acid synthesis and is effective against various gram-positive and gram-negative bacteria.
  • Exhibits antifungal and antibacterial activities
  • Used to treat bacterial infections, acne, and scalp conditions
  • Targets bacterial folic acid synthesis
  • Effective against various gram-positive and gram-negative bacteria
  • Inhibits the synthesis of both dihydro-folic acid and para-aminobenzoic acid
  • Can be utilized for studying ocular infections
Catalog No. 50-004-19922