Learn More
Medchemexpress LLC Sulfadimethoxine-d4 | 1020719-80-5 | ≥98.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0337S1MG
Encompass_Preferred
Sulfadimethoxine-d4 is a deuterium-labeled version of Sulfadimethoxine, a sulfonamide antibiotic effective against various infections, including those affecting the respiratory and urinary tracts, as well as enteric and soft tissue infections. This compound is intended for research use only and is not sold to patients.
  • Can be utilized as a tracer
  • Can be utilized as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-004-06704