Learn More
Medchemexpress LLC Sulfadimethoxine-d6 | 73068-02-7 | 99.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0337S15MG
Encompass_Preferred
Sulfadimethoxine-d6 is the deuterium-labeled form of Sulfadimethoxine, a sulfonamide antibiotic used to treat various infections. This compound is intended for research use only.
  • Used as a tracer or internal standard for quantitative analysis
  • Suitable for methods such as NMR, GC-MS, or LC-MS
  • Isotope-labeled compound
  • Internal standard for clinical mass spectrometry, food, pesticides, and veterinary medicine
  • Stable isotope-labeled inhibitor for in vivo cell experiments
  • Used in therapeutic drug monitoring (TDM)
Catalog No. 50-004-07256