Learn More
Medchemexpress LLC Sulfadimethoxine sodium | 1037-50-9 | 99.4% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0337A10MM/1ML
Encompass_Preferred
Sulfadimethoxine sodium is a sulfonamide antibiotic primarily used in various infection studies, including those focused on urinary tract infections. It has been observed to cause a displacement of Thiopental in plasma protein binding in rats.
  • Increases the apparent dissociation constant of Thiopental to serum protein by equilibrium dialysis
  • Displaces Thiopental in plasma protein binding in rats, significantly increasing the free fraction of thiopental
Catalog No. 50-004-07274