Learn More
Medchemexpress LLC Sulfamerazine | 127-79-7 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB051210MM/1ML
Encompass_Preferred
Sulfamerazine (RP-2632) is a sulfonamide antibacterial. It is the monomethyl derivative of sulfadiazine, specifically 2-sulfanilamido-4-methylpyrimidine. This sulfonamide drug inhibits bacterial synthesis of dihydrofolic acid by competing with para-aminobenzoic acid (PABA) for binding to dihydropteroate synthesizes.
  • Functions as a sulfonamide antibacterial.
  • Inhibits bacterial synthesis of dihydrofolic acid.
  • Competes with para-aminobenzoic acid (PABA) for binding.
  • Monomethyl derivative of sulfadiazine.
Catalog No. 50-004-05551