Learn More
Apexbio Technology LLC Sulfameter 651-06-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B20385.1
Encompass_Preferred
Sulfameter (651-06-9) is a small-molecule inhibitor targeting bacterial dihydropteroate synthase It is designed to competitively inhibit this enzyme thereby interfering with folate biosynthesis essential for bacterial growth Sulfameter exerts its biological activity primarily through competitive inhibition of bacterial dihydropteroate synthase In biochemical assays Sulfameter demonstrates inhibitory activity with reported IC50 values typically within the micromolar range depending on experimental conditions and bacterial strains tested Based on these pharmacological properties Sulfameter holds research potential in studies of bacterial growth inhibition resistance mechanisms mechanism-of-action assays and evaluations of antibacterial susceptibility
Catalog No. 50-000-93018