Learn More
Medchemexpress LLC Sulfameter | 651-06-9 | 99.9% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB021310G
Encompass_Preferred
Sulfameter (Sulfametoxydiazine) is an orally active, long-acting sulfonamide antibiotic that demonstrates activity against both Gram-positive and Gram-negative bacteria. It is suitable for research on diseases like respiratory and urinary tract infections. In in vitro studies, it forms a complex with silver, showing a more pronounced effect on Gram-negative bacteria.
  • Active against Gram-positive and Gram-negative bacteria
  • Suitable for research into respiratory and urinary tract infections
  • Forms a complex with silver to demonstrate activity against bacteria
Catalog No. 50-004-05747