Learn More
Cayman Chemical SulfamethazIn-d4 5mg
SDP
Supplier:  Cayman Chemical 365755 MG
Encompass_Preferred
An internal standard for the quantification of sulfamethazine by GC- or LC-MS
Catalog No. 50-290-9279