Learn More
Medchemexpress LLC Sulfamethazine-d4 (sulfadimidine-d4) | 1020719-82-7 | MFCD03701230 | 99.9% | 282.35 g/mol | C12H10D4N4O2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0035S10MG
Encompass_Preferred
Sulfamethazine-d4 is a deuterium-labeled analogue of sulfamethazine supplied as a solid for analytical and research use. It is used primarily as an isotopically labeled internal standard or tracer for quantitative analyses by LC-MS, GC-MS, and NMR. The material is provided at high purity with defined storage recommendations to ensure stability in analytical workflows.
  • Deuterium-labeled internal standard for quantitative assays.
  • High purity suitable for reference and analytical use.
  • White to off-white solid form, stable when stored at -20°C.
  • Compatible with LC-MS, GC-MS, and NMR methods.
  • Includes common synonyms for easy cross-referencing.
Catalog No. 50-002-97383