Learn More
Medchemexpress LLC Sulfamethazine sodium | 1981-58-4 | 99.9% | C12H13N4NaO2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0035A10MM/1ML
Encompass_Preferred
Sulfamethazine sodium is an antimicrobial that can cross the blood-brain barrier. It is commonly used to treat and prevent various animal diseases, such as gastrointestinal and respiratory tract infections. A maximum residue level of 100 μg/kg for Sulfamethazine sodium in animal products has been established by China and the European Commission.
  • Antimicrobial with blood-brain barrier permeability
  • Treats and prevents various animal diseases
  • Effective against gastrointestinal and respiratory tract infections
Catalog No. 50-004-32763