Learn More
Accela Chembio Inc Sulfamoyl Chloride | 25g | 7778-42-9 | MFCD11519990 | 95+% | Shelf Life: 720 Days | Moisture Sensitive/store In Freezer(-20) Upon Receipt/air Sensitive
Supplier:  Accela Chembio Inc SY00641525G
Encompass_Preferred
Sulfamoyl Chloride | 25g | 7778-42-9 | MFCD11519990 | 95+% | Shelf Life: 720 Days | Moisture Sensitive/store In Freezer(-20) Upon Receipt/air Sensitive
Catalog No. 50-354-509