Learn More
Apexbio Technology LLC Sulfanilamide,10g CAS# 63-74-1
SDP
Supplier:  Apexbio Technology LLC B204110000
Encompass_Preferred
Sulfanilamide is a competitive inhibitor for bacterial enzyme dihydropteroate synthetase with IC50 of 320 μM. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0400