Learn More
Cayman Chemical Sulfaphenazole 250mg
SDP
Supplier:  Cayman Chemical 14844250 MG
Encompass_Preferred
Selective inhibitor of CYP2C9 (Ki = 0.3 μM); inhibit endothelium-derived hyperpolarizing factor synthase, a CYP450 isozyme in the porcine coronary artery homologous to CYP2C8/9 that generates reactive oxygen species in coronary endothelial cells and modulates vascular tone and homeostasis
Catalog No. 50-296-8528