Learn More
Apexbio Technology LLC Sulfaphenazole 526-08-9 250mg
SDP
Supplier:  Apexbio Technology LLC C4131250
Encompass_Preferred
Sulfaphenazole (CAS 526-08-9) is a small-molecule inhibitor targeting cytochrome P450 2C9 (CYP2C9) It is designed to selectively inhibit CYP2C9-mediated drug metabolism thereby informing metabolic pathway studies and investigation of drug-drug interactions Sulfaphenazole exerts its biological activity primarily through competitive inhibition of CYP2C9 In in vitro assays sulfaphenazole demonstrates potent inhibition with a reported Ki value of approximately 0 3 M for human CYP2C9 Based on these pharmacological properties sulfaphenazole holds research potential in evaluating CYP2C9-mediated metabolic processes and associated drug interactions
Catalog No. 50-000-91250