Learn More
Medchemexpress LLC Sulfaquinoxaline | 59-40-5 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1282100MG
Encompass_Preferred
Sulfaquinoxaline is an antimicrobial agent used in veterinary medicine. It demonstrates activity against a wide range of both Gram-negative and Gram-positive bacteria. This product is employed for the prevention of coccidiosis and various bacterial infections.
  • Antimicrobial for veterinary use.
  • Active against a broad spectrum of gram-negative bacteria.
  • Active against a broad spectrum of gram-positive bacteria.
  • Used to prevent coccidiosis.
  • Used to prevent bacterial infections.
Catalog No. 50-004-12963