Learn More
Medchemexpress LLC Sulfaquinoxaline (sodium salt) | 967-80-6 | 99.7% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB1282A10MM/1ML
Encompass_Preferred
Sulfaquinoxaline sodium salt is an antimicrobial used in veterinary medicine, effective against a broad range of Gram-negative and Gram-positive bacteria. This substance is used to prevent coccidiosis and bacterial infections. It is for research use only.
  • Antimicrobial for veterinary use
  • Activity against a broad spectrum of Gram-negative and Gram-positive bacteria
  • Used to prevent coccidiosis and bacterial infections
Catalog No. 50-004-12982