Learn More
Selleck Chemical LLC Sulfasalazine S1576-500mg
SDP
Supplier:  Selleck Chemical LLC S1576500MG
Encompass_Preferred
Sulfasalazine is a sulfa derivative of mesalazine used as an anti-inflammatory agent to treat bowel disease and rheumatoid arthritis Sulfasalazine is a potent and specific inhibitor of nuclear factor kappa B (NF- B) TGF- and COX-2 Sulfasalazine induces ferroptosis apoptosis and autophagy
Catalog No. 50-311-0096