Learn More
Hach Company Sulfate TNTplus Vial Test, LR (40-150 mg/L SO₄), 25 Tests
Supplier:  Hach Company TNT864
Encompass
Sulfate TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Sulfate by the Turbidimetric Method.
Catalog No. 50-199-8149