Learn More
Apexbio Technology LLC Sulfatinib 1308672-74-3 1mg
SDP
Supplier:  Apexbio Technology LLC A47421
Encompass_Preferred
Sulfatinib (CAS 1308672-74-3) is a small-molecule tyrosine kinase inhibitor that selectively targets vascular endothelial growth factor receptors (VEGFR) 1-3 fibroblast growth factor receptor 1 (FGFR1) and colony-stimulating factor-1 receptor (CSF1R) By inhibiting these signaling pathways sulfatinib disrupts tumor angiogenesis and modulates the tumor microenvironment The compound is currently being evaluated in clinical studies for the treatment of advanced neuroendocrine tumors including a phase III trial in patients with extrapancreatic neuroendocrine tumors supporting its application in cancer research focused on anti-angiogenic therapy
Catalog No. 50-000-95100