Learn More
Medchemexpress LLC HY-B0323 100mg Medchemexpress, Sulfisoxazole CAS:127-69-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0323100MG
Encompass_Preferred
Medchemexpress, HY-B0323 100mg Sulfisoxazole CAS:127-69-5 Sulfisoxazole (Sulfafurazole), an endothelin receptor antagonist, is a sulfonamide antibacterial with an oxazole substituent. Sulfisoxazole inhibits breast cancer exosome release by targeting endothelin receptor A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2468