Learn More
Medchemexpress LLC Sulfo-ara-F-NMN | 1374663-29-2 | 99.4% | 352.28 | 50 MG
SDP
Supplier:  Medchemexpress LLC 50MGHY129522
Encompass_Preferred
Sulfo-ara-F-NMN (CZ-48) is a mimetic of nicotinamide mononucleotide (NMN) that selectively activates SARM1 and inhibits CD38 (IC50 around 10 μM). It induces intracellular cyclic ADP-ribose (cADPR) production and is intended for research use only.
  • Selectively activates SARM1
  • Inhibits CD38
  • Induces intracellular cyclic ADP-ribose (cADPR) production
  • Appears as a white to light yellow solid
  • Store at -20°C under nitrogen
  • Compound is unstable in solutions; freshly prepared is recommended
Catalog No. 50-003-65981