Learn More
Medchemexpress LLC Sulfo-Cy3(Me)COOH | 1121756-11-3 | 95.0% | 616.75 g·mol⁻¹ | C30H36N2O8S2 | 10 MG

Supplier: Medchemexpress LLC HYD136510MG
Sulfo-Cy3(Me)COOH is a sulfonated cyanine3 derivative supplied as a green to dark green solid. It is an orange-fluorescent labeling reagent for proteins and nucleic acids, with excitation at ~554 nm and emission at ~568 nm. The compound is water-soluble and is provided at 10 mg with high purity, intended for conjugation and fluorescence assay applications and requiring protection from light.
- Orange-fluorescent label: Ex = 554 nm, Em = 568 nm.
- High purity suitable for conjugation: ~95.0%.
- Water-soluble: H2O solubility 125 mg/mL (ultrasonic may be required).
- Appearance: solid, green to dark green.
- Storage: 4°C protected from light; in solvent -80°C (6 months) or -20°C (1 month).
- Typical applications: labeling of proteins and nucleic acids, fluorescence assays, conjugation chemistry.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.